Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLK1
Entrez Gene ID
9874
Gene Name
tousled like kinase 1
Gene Aliases
PKU-beta
Chromosome Location
Chr.2: 170990823 - 171231314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136554.1 NP_001130026.1 15 - 16 1544 123
NM_001136555.1 NP_001130027.1 13 - 14 1315 123
NM_012290.4 NP_036422.3 15 - 16 1985 123
XM_011512238.2 XP_011510540.1 14 - 15 1836 123
XM_017005421.1 XP_016860910.1 16 - 17 2094 123
XM_017005422.1 XP_016860911.1 14 - 15 1906 123
GenBank mRNA AB004885.1 - 16 - 17 1788 123
AF162666.1 - 15 - 16 1666 123
AF246219.1 - 8 - 9 871 123
AK090779.1 - 15 - 16 1544 123
AK091975.1 - 16 - 17 2243 123
AK292508.1 - 15 - 16 1626 123
AK301857.1 - 13 - 14 1315 123
BC032657.1 - 15 - 16 1546 123
D50927.2 - 15 - 16 1618 123

View Full Product Details