Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLK1
Entrez Gene ID
9874
Gene Name
tousled like kinase 1
Gene Aliases
PKU-beta
Chromosome Location
Chr.2: 170990823 - 171231314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136554.1 NP_001130026.1 9 - 10 877 138
NM_001136555.1 NP_001130027.1 7 - 8 648 138
NM_012290.4 NP_036422.3 9 - 10 1318 138
XM_011512238.2 XP_011510540.1 9 - 10 1358 138
XM_017005421.1 XP_016860910.1 10 - 11 1427 138
XM_017005422.1 XP_016860911.1 8 - 9 1239 138
GenBank mRNA AB004885.1 - 10 - 11 138
AF162666.1 - 9 - 10 999 138
AF246219.1 - 2 - 3 204 138
AK090779.1 - 9 - 10 877 138
AK091975.1 - 9 - 10 1318 138
AK292508.1 - 9 - 10 959 138
AK301857.1 - 7 - 8 648 138
BC032657.1 - 9 - 10 879 138
BX537631.1 - 2 - 3 1481 138
D50927.2 - 9 - 10 951 138

View Full Product Details