Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECAP2
Entrez Gene ID
55707
Gene Name
NECAP endocytosis associated 2
Gene Aliases
-
Chromosome Location
Chr.1: 16440672 - 16460090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145277.1 NP_001138749.1 6 - 7 756 53
XM_011541767.2 XP_011540069.1 7 - 8 828 53
GenBank mRNA AK023545.1 - 6 - 7 692 53

View Full Product Details