Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
Chromosome Location
Chr.1: 40300487 - 40317653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001852.3 NP_001843.1 26 - 27 1444 78
XM_006710365.3 XP_006710428.1 27 - 28 1552 78
XM_011540715.2 XP_011539017.1 26 - 27 2032 78
XM_011540716.2 XP_011539018.1 25 - 26 1512 78
XM_011540717.2 XP_011539019.1 25 - 26 2360 78
XM_017000332.1 XP_016855821.1 26 - 27 1498 78
XM_017000333.1 XP_016855822.1 25 - 26 1498 78
GenBank mRNA BC041479.1 - 25 - 26 1677 78
BC136326.1 - 26 - 27 1437 78
BC136327.1 - 26 - 27 1437 78
M95610.1 - 26 - 27 1333 78

View Full Product Details