Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
Chromosome Location
Chr.1: 40300487 - 40317653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001852.3 NP_001843.1 4 - 5 316 107
XM_006710365.3 XP_006710428.1 5 - 6 424 107
XM_011540715.2 XP_011539017.1 4 - 5 892 107
XM_011540716.2 XP_011539018.1 3 - 4 372 107
XM_011540717.2 XP_011539019.1 4 - 5 889 107
XM_017000332.1 XP_016855821.1 4 - 5 358 107
XM_017000333.1 XP_016855822.1 3 - 4 370 107
GenBank mRNA BC041479.1 - 4 - 5 206 107
BC136326.1 - 4 - 5 309 107
BC136327.1 - 4 - 5 309 107
M95610.1 - 4 - 5 205 107

View Full Product Details