The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001164410.2 | NP_001157882.1 | 9 - 10 | 733 | 74 | |
NM_004935.3 | NP_004926.1 | 10 - 11 | 829 | 74 | ||
GenBank mRNA | AK026533.1 | - | 10 - 11 | 750 | 74 | |
AY049778.1 | - | 10 - 11 | 712 | 74 | ||
AY927560.1 | - | LIT3563 AY927560 |
5 - 6 | 751 | 74 | |
BC005115.1 | - | 10 - 11 | 737 | 74 | ||
BT006680.1 | - | 10 - 11 | 712 | 74 | ||
CA429180.1 | - | 2 - 3 | 74 | |||
CR457041.1 | - | 10 - 11 | 712 | 74 | ||
DQ411039.1 | - | 9 - 10 | 616 | 74 | ||
KU177968.1 | - | 10 - 11 | 712 | 74 | ||
KU177969.1 | - | 9 - 10 | 616 | 74 | ||
L04658.1 | - | 10 - 11 | 724 | 74 | ||
X66364.1 | - | 10 - 11 | 736 | 74 |