Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MX1
Entrez Gene ID
4599
Gene Name
MX dynamin like GTPase 1
Gene Aliases
IFI-78K, IFI78, MX, MxA
Chromosome Location
Chr.21: 41420329 - 41459214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144925.2 NP_001138397.1 17 - 18 2492 109
NM_001178046.2 NP_001171517.1 13 - 14 1876 109
NM_002462.4 NP_002453.2 15 - 16 2051 109
XM_005260978.4 XP_005261035.1 15 - 16 1967 109
XM_005260979.2 XP_005261036.1 15 - 16 2160 109
XM_005260980.2 XP_005261037.1 15 - 16 1932 109
XM_005260981.2 XP_005261038.1 14 - 15 1812 109
XM_005260982.2 XP_005261039.1 14 - 15 1797 109
XM_011529568.2 XP_011527870.1 19 - 20 2501 109
XM_017028349.1 XP_016883838.1 17 - 18 2511 109
XM_017028350.1 XP_016883839.1 15 - 16 2179 109
XM_017028351.1 XP_016883840.1 14 - 15 1835 109
XM_017028352.1 XP_016883841.1 14 - 15 1808 109
GenBank mRNA AK093008.1 - 11 - 12 1799 109
AK096355.1 - 17 - 18 2535 109
AK315465.1 - 13 - 14 1733 109
BC014222.2 - 10 - 11 1260 109
BC032602.1 - 14 - 15 1801 109
M30817.1 - 13 - 14 1720 109
M33882.1 - 15 - 16 1855 109

View Full Product Details