Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCRA
Entrez Gene ID
171558
Gene Name
pre T-cell antigen receptor alpha
Gene Aliases
PT-ALPHA, PTA
Chromosome Location
Chr.6: 42915989 - 42925838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243169.1 NP_001230098.1 1 - 2 139 81
GenBank mRNA AF165312.1 - 1 - 2 58 81
AK303523.1 - 1 - 2 65 81

View Full Product Details