Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC11A2
Entrez Gene ID
4891
Gene Name
solute carrier family 11 member 2
Gene Aliases
AHMIO1, DCT1, DMT1, NRAMP2
Chromosome Location
Chr.12: 50952263 - 51028335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000617.2 NP_000608.1 14 - 15 1568 63
NM_001174125.1 NP_001167596.1 14 - 15 1560 63
NM_001174126.1 NP_001167597.1 14 - 15 1568 63
NM_001174127.1 NP_001167598.1 14 - 15 1563 63
NM_001174128.1 NP_001167599.1 14 - 15 1563 63
NM_001174129.1 NP_001167600.1 14 - 15 1828 63
NM_001174130.1 NP_001167601.1 13 - 14 1543 63
NR_033421.1 14 - 15 1488 63
NR_033422.1 14 - 15 1563 63
XM_005268911.3 XP_005268968.1 14 - 15 1620 63
XM_005268912.4 XP_005268969.1 13 - 14 1745 63
XM_011538404.2 XP_011536706.1 14 - 15 1944 63
XM_011538405.2 XP_011536707.1 14 - 15 1552 63
XM_017019355.1 XP_016874844.1 14 - 15 1828 63
XM_017019356.1 XP_016874845.1 14 - 15 1758 63
GenBank mRNA AB004857.1 - 14 - 15 1511 63
AB062284.1 - 14 - 15 1485 63
AF046997.1 - 14 - 15 1521 63
AF064484.1 - 14 - 15 1511 63
AJ493662.1 - 14 - 15 1560 63
AK094735.1 - 14 - 15 1828 63
AK128640.1 - 14 - 15 1460 63
AK296445.1 - 13 - 14 1522 63
AK315987.1 - 14 - 15 1474 63
AK316507.1 - 13 - 14 1499 63
AL832886.1 - 4 - 5 894 63
BC002592.2 - 14 - 15 1515 63
BC100014.1 - 14 - 15 1536 63
CD365137.1 - 4 - 5 63
KT455015.1 - 14 - 15 1524 63

View Full Product Details