The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022437.2 | NP_071882.1 | 10 - 11 | 1576 | 80 |
XM_005264483.3 | XP_005264540.1 | 10 - 11 | 1688 | 80 | |
XM_011533029.2 | XP_011531331.1 | 10 - 11 | 2143 | 80 | |
XM_011533030.2 | XP_011531332.1 | 10 - 11 | 1985 | 80 | |
GenBank mRNA | AF320294.1 | - | 10 - 11 | 1486 | 80 |
AF324494.1 | - | 10 - 11 | 1576 | 80 | |
BC113657.1 | - | 10 - 11 | 1522 | 80 | |
BC143564.1 | - | 10 - 11 | 1519 | 80 |