The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152434.2 | NP_689647.2 | 5 - 6 | 595 | 117 |
XM_011542620.2 | XP_011540922.1 | 5 - 6 | 593 | 117 | |
XM_017017239.1 | XP_016872728.1 | 5 - 6 | 566 | 117 | |
GenBank mRNA | AK056905.1 | - | 3 - 4 | 247 | 117 |
AK126881.1 | - | 5 - 6 | 588 | 117 | |
AK293001.1 | - | 5 - 6 | 530 | 117 | |
BC070142.1 | - | 5 - 6 | 574 | 117 | |
BC107418.1 | - | 5 - 6 | 578 | 117 | |
BC110439.1 | - | 2 - 3 | 208 | 117 | |
BC110440.1 | - | 2 - 3 | 208 | 117 | |
BC110441.1 | - | 2 - 3 | 208 | 117 | |
BC110442.1 | - | 2 - 3 | 208 | 117 | |
BC118669.1 | - | 2 - 3 | 211 | 117 | |
BX647832.1 | - | 5 - 6 | 569 | 117 | |
BX648704.1 | - | 5 - 6 | 574 | 117 | |
BX649022.1 | - | 5 - 6 | 565 | 117 |