Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Chromosome Location
Chr.9: 86026141 - 86100201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016548.3 NP_057632.2 8 - 9 1226 117
NM_177937.2 NP_808800.1 8 - 9 1218 117
GenBank mRNA AF236056.1 - 8 - 9 1167 117
AK027717.1 - 8 - 9 1189 117
AK075542.1 - 8 - 9 1160 117
AK075557.1 - 8 - 9 1130 117
AK092943.1 - 9 - 10 1106 117
AY358593.1 - 8 - 9 1131 117
BC001740.1 - 8 - 9 1141 117
CR457201.1 - 7 - 8 1017 117
EU794616.1 - 8 - 9 1185 117

View Full Product Details