Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATXN7L3
Entrez Gene ID
56970
Gene Name
ataxin 7 like 3
Gene Aliases
-
Chromosome Location
Chr.17: 44191805 - 44199976 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098833.1 NP_001092303.1 1 - 2 148 61
NM_020218.1 NP_064603.1 1 - 2 148 61
XM_017024883.1 XP_016880372.1 2 - 3 323 61
XM_017024884.1 XP_016880373.1 1 - 2 366 61
XM_017024885.1 XP_016880374.1 2 - 3 323 61
XM_017024886.1 XP_016880375.1 2 - 3 323 61
GenBank mRNA AK126117.1 - 1 - 2 366 61
BC037418.1 - 1 - 2 55 61
BC113595.1 - 1 - 2 148 61
BC126113.1 - 1 - 2 148 61

View Full Product Details