The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007202.3 | NP_009133.2 | 2 - 3 | 312 | 98 |
XM_006721432.3 | XP_006721495.1 | 2 - 3 | 307 | 98 | |
XM_017024071.1 | XP_016879560.1 | 2 - 3 | 307 | 98 | |
GenBank mRNA | AF037439.1 | - | 2 - 3 | 272 | 98 |
AK308297.1 | - | 2 - 3 | 159 | 98 | |
AK312438.1 | - | 2 - 3 | 265 | 98 | |
BC017055.1 | - | 2 - 3 | 268 | 98 | |
BP229845.1 | - | 2 - 3 | 312 | 98 | |
DQ270159.1 | - | 2 - 3 | 135 | 98 | |
KU178546.1 | - | 2 - 3 | 135 | 98 | |
KU178547.1 | - | 2 - 3 | 135 | 98 |