The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007202.3 | NP_009133.2 | 6 - 7 | 1237 | 69 |
XM_006721432.3 | XP_006721495.1 | 6 - 7 | 1232 | 69 | |
XM_017024071.1 | XP_016879560.1 | 6 - 7 | 1232 | 69 | |
GenBank mRNA | AF037439.1 | - | 6 - 7 | 1197 | 69 |
AK312438.1 | - | 6 - 7 | 1190 | 69 | |
BC017055.1 | - | 6 - 7 | 1193 | 69 | |
DQ270159.1 | - | 6 - 7 | 1060 | 69 | |
KU178546.1 | - | 6 - 7 | 1060 | 69 | |
KU178547.1 | - | 6 - 7 | 1060 | 69 | |
KU178548.1 | - | 5 - 6 | 877 | 69 |