The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003934.1 | NP_003925.1 | 9 - 10 | 884 | 66 |
XM_005272232.1 | XP_005272289.1 | 9 - 10 | 890 | 66 | |
XM_006717313.2 | XP_006717376.1 | 10 - 11 | 1735 | 66 | |
XM_011519172.2 | XP_011517474.1 | 9 - 10 | 890 | 66 | |
XM_011519173.2 | XP_011517475.1 | 7 - 8 | 1142 | 66 | |
XM_011519174.2 | XP_011517476.1 | 11 - 12 | 1980 | 66 | |
XM_017015269.1 | XP_016870758.1 | 10 - 11 | 1735 | 66 | |
XM_017015270.1 | XP_016870759.1 | 9 - 10 | 1677 | 66 | |
XM_017015271.1 | XP_016870760.1 | 10 - 11 | 1922 | 66 | |
XM_017015272.1 | XP_016870761.1 | 7 - 8 | 705 | 66 | |
XM_017015273.1 | XP_016870762.1 | 9 - 10 | 1677 | 66 | |
GenBank mRNA | AK295155.1 | - | 9 - 10 | 884 | 66 |
AL537190.3 | - | 9 - 10 | 855 | 67 | |
BC001325.1 | - | 1 - 2 | 44 | 66 | |
BC007874.2 | - | 10 - 11 | 954 | 66 | |
BC137338.1 | - | 9 - 10 | 881 | 66 | |
BC137340.1 | - | 9 - 10 | 881 | 66 | |
U69127.1 | - | 9 - 10 | 860 | 66 |