Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOA2
Entrez Gene ID
10499
Gene Name
nuclear receptor coactivator 2
Gene Aliases
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
Chromosome Location
Chr.8: 70109762 - 70405390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321703.1 NP_001308632.1 22 - 23 4670 93
NM_001321707.1 NP_001308636.1 22 - 23 4516 93
NM_001321711.1 NP_001308640.1 21 - 22 4391 93
NM_001321712.1 NP_001308641.1 21 - 22 4308 93
NM_001321713.1 NP_001308642.1 21 - 22 4411 93
NM_006540.3 NP_006531.1 22 - 23 4598 93
XM_017012961.1 XP_016868450.1 22 - 23 4792 93
XM_017012962.1 XP_016868451.1 22 - 23 4816 93
XM_017012963.1 XP_016868452.1 22 - 23 6029 93
XM_017012964.1 XP_016868453.1 22 - 23 5084 93
XM_017012965.1 XP_016868454.1 22 - 23 4749 93
XM_017012966.1 XP_016868455.1 23 - 24 5013 93
XM_017012967.1 XP_016868456.1 21 - 22 4727 93
XM_017012968.1 XP_016868457.1 23 - 24 4814 93
XM_017012969.1 XP_016868458.1 21 - 22 4845 93
XM_017012970.1 XP_016868459.1 21 - 22 11264 93
XM_017012971.1 XP_016868460.1 21 - 22 4976 93
XM_017012972.1 XP_016868461.1 18 - 19 4307 93
GenBank mRNA AK298528.1 - 10 - 11 1683 93
BC114383.1 - 21 - 22 4465 93
X97674.1 - 22 - 23 4543 93

View Full Product Details