Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOA2
Entrez Gene ID
10499
Gene Name
nuclear receptor coactivator 2
Gene Aliases
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
Chromosome Location
Chr.8: 70109762 - 70405390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321703.1 NP_001308632.1 5 - 6 648 87
NM_001321707.1 NP_001308636.1 5 - 6 494 87
NM_001321711.1 NP_001308640.1 5 - 6 576 87
NM_001321712.1 NP_001308641.1 5 - 6 493 87
NM_006540.3 NP_006531.1 5 - 6 576 87
XM_017012961.1 XP_016868450.1 5 - 6 563 87
XM_017012962.1 XP_016868451.1 5 - 6 587 87
XM_017012963.1 XP_016868452.1 5 - 6 1800 87
XM_017012964.1 XP_016868453.1 5 - 6 855 87
XM_017012965.1 XP_016868454.1 5 - 6 520 87
XM_017012966.1 XP_016868455.1 6 - 7 784 87
XM_017012967.1 XP_016868456.1 4 - 5 498 87
XM_017012968.1 XP_016868457.1 6 - 7 585 87
XM_017012969.1 XP_016868458.1 4 - 5 616 87
XM_017012970.1 XP_016868459.1 4 - 5 7035 87
XM_017012971.1 XP_016868460.1 4 - 5 747 87
XM_017012973.1 XP_016868462.1 5 - 6 563 87
GenBank mRNA AK308426.1 - 5 - 6 541 87
BC114383.1 - 4 - 5 443 87
CK002637.1 - 5 - 6 494 87
X97674.1 - 5 - 6 521 87

View Full Product Details