Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NCOA2
- Entrez Gene ID
-
10499
- Gene Name
- nuclear receptor coactivator 2
- Gene Aliases
-
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
- Chromosome Location
-
Chr.8: 70109762 - 70405390 on Build GRCh38
View Full Product Details