Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLE1
Entrez Gene ID
7088
Gene Name
transducin like enhancer of split 1
Gene Aliases
ESG, ESG1, GRG1
Chromosome Location
Chr.9: 81583683 - 81689535 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303103.1 NP_001290032.1 18 - 19 3449 79
NM_001303104.1 NP_001290033.1 18 - 19 3374 79
NM_005077.4 NP_005068.2 18 - 19 3419 79
XM_005252151.1 XP_005252208.1 19 - 20 2465 79
XM_005252152.1 XP_005252209.1 19 - 20 2462 79
XM_005252153.1 XP_005252210.1 19 - 20 2435 79
XM_005252154.1 XP_005252211.1 19 - 20 2432 79
XM_005252156.2 XP_005252213.1 18 - 19 2360 79
XM_005252162.1 XP_005252219.1 17 - 18 2213 79
XM_005252163.2 XP_005252220.1 16 - 17 2111 79
XM_006717258.1 XP_006717321.1 19 - 20 2462 79
XM_006717259.3 XP_006717322.1 17 - 18 2148 79
XM_006717260.1 XP_006717323.1 17 - 18 2243 79
XM_006717261.2 XP_006717324.1 17 - 18 2177 79
XM_006717262.1 XP_006717325.1 18 - 19 2330 79
XM_006717263.1 XP_006717326.1 18 - 19 2327 79
XM_011518951.2 XP_011517253.1 18 - 19 2330 79
XM_017015064.1 XP_016870553.1 18 - 19 2360 79
XM_017015065.1 XP_016870554.1 18 - 19 2357 79
XM_017015066.1 XP_016870555.1 16 - 17 2103 79
GenBank mRNA AB209854.1 - 17 - 18 6406 79
AK290860.1 - 18 - 19 2181 79
AK293606.1 - 18 - 19 2162 79
AK304565.1 - 14 - 15 1636 79
BC010100.2 - 18 - 19 2565 79
BC015747.1 - 18 - 19 2275 79
M99435.1 - 18 - 19 2149 79

View Full Product Details