Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLE1
Entrez Gene ID
7088
Gene Name
transducin like enhancer of split 1
Gene Aliases
ESG, ESG1, GRG1
Chromosome Location
Chr.9: 81583683 - 81689535 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303103.1 NP_001290032.1 2 - 3 1421 88
NM_001303104.1 NP_001290033.1 2 - 3 1421 88
NM_005077.4 NP_005068.2 2 - 3 1421 88
XM_005252151.1 XP_005252208.1 2 - 3 335 88
XM_005252152.1 XP_005252209.1 2 - 3 335 88
XM_005252153.1 XP_005252210.1 2 - 3 335 88
XM_005252154.1 XP_005252211.1 2 - 3 335 88
XM_005252156.2 XP_005252213.1 2 - 3 335 88
XM_005252162.1 XP_005252219.1 2 - 3 335 88
XM_005252163.2 XP_005252220.1 2 - 3 335 88
XM_006717258.1 XP_006717321.1 2 - 3 335 88
XM_006717259.3 XP_006717322.1 2 - 3 186 88
XM_006717260.1 XP_006717323.1 2 - 3 335 88
XM_006717262.1 XP_006717325.1 2 - 3 335 88
XM_006717263.1 XP_006717326.1 2 - 3 335 88
XM_011518951.2 XP_011517253.1 2 - 3 335 88
XM_017015064.1 XP_016870553.1 2 - 3 335 88
XM_017015065.1 XP_016870554.1 2 - 3 335 88
XM_017015066.1 XP_016870555.1 2 - 3 243 88
GenBank mRNA AB209854.1 - 2 - 3 1421 88
AK290860.1 - 2 - 3 183 88
AK293606.1 - 2 - 3 209 88
AK304565.1 - 2 - 3 244 88
BC010100.2 - 2 - 3 567 88
BC015747.1 - 2 - 3 277 88
M99435.1 - 2 - 3 151 88

View Full Product Details