The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270940.1 | NP_001257869.1 | 14 - 15 | 2453 | 95 |
NM_015171.3 | NP_055986.1 | 13 - 14 | 2317 | 95 | |
XM_005255197.3 | XP_005255254.1 | 11 - 12 | 1651 | 95 | |
XM_005255198.1 | XP_005255255.1 | 9 - 10 | 1800 | 95 | |
XM_005255199.2 | XP_005255256.1 | 9 - 10 | 994 | 95 | |
XM_011545772.2 | XP_011544074.1 | 13 - 14 | 1986 | 95 | |
GenBank mRNA | AB002368.3 | - | 13 - 14 | 2268 | 95 |
AK026172.1 | - | 10 - 11 | 1461 | 95 | |
AK123846.1 | - | 14 - 15 | 2085 | 95 | |
AY026388.1 | - | 13 - 14 | 1790 | 95 | |
BC004403.1 | - | 7 - 8 | 1057 | 95 | |
BC014071.2 | - | 7 - 8 | 1057 | 95 | |
BC078674.1 | - | 9 - 10 | 1262 | 95 | |
BC108286.1 | - | 9 - 10 | 1216 | 95 | |
BC130304.1 | - | 13 - 14 | 1790 | 95 | |
BC144189.1 | - | 13 - 14 | 1790 | 95 |