The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079.3 | NP_001070.2 | 3 - 4 | 609 | 108 |
XM_017004867.1 | XP_016860356.1 | 5 - 6 | 2010 | 108 | |
XM_017004868.1 | XP_016860357.1 | 5 - 6 | 2010 | 108 | |
XM_017004869.1 | XP_016860358.1 | 5 - 6 | 2010 | 108 | |
XM_017004870.1 | XP_016860359.1 | 5 - 6 | 2010 | 108 | |
GenBank mRNA | AK303341.1 | - | 3 - 4 | 601 | 108 |
AK310628.1 | - | 3 - 4 | 595 | 108 | |
AK310637.1 | - | 3 - 4 | 601 | 108 | |
BC053878.1 | - | 3 - 4 | 585 | 108 | |
L05148.1 | - | 3 - 4 | 611 | 108 |