Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf81
Entrez Gene ID
388963
Gene Name
chromosome 2 open reading frame 81
Gene Aliases
hCG40743
Chromosome Location
Chr.2: 74414176 - 74421630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145054.1 NP_001138526.1 1 - 2 335 98

View Full Product Details