The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287491.1 | NP_001274420.1 | 8 - 9 | 3130 | 83 |
XM_005264187.3 | XP_005264244.1 | 9 - 10 | 3650 | 83 | |
XM_011532686.2 | XP_011530988.1 | 8 - 9 | 3593 | 83 | |
XM_017003567.1 | XP_016859056.1 | 8 - 9 | 2962 | 83 | |
GenBank mRNA | BC022243.1 | - | 5 - 6 | 586 | 83 |
HQ220209.1 | - | 8 - 9 | 3130 | 83 |