Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNKS
Entrez Gene ID
8658
Gene Name
tankyrase
Gene Aliases
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5
Chromosome Location
Chr.8: 9555229 - 9782346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003747.2 NP_003738.2 11 - 12 1753 96
XM_006716263.3 XP_006716326.1 11 - 12 2459 96
XM_011543845.2 XP_011542147.1 11 - 12 2459 96
XM_011543846.2 XP_011542148.1 11 - 12 2459 96
XM_011543847.2 XP_011542149.1 11 - 12 2458 96
GenBank mRNA AB209339.1 - 10 - 11 933 96
AF082556.1 - 11 - 12 1753 96
AF082557.1 - 10 - 11 1049 96
AK302534.1 - 11 - 12 1427 96
BC098394.1 - 11 - 12 1776 96

View Full Product Details