The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003747.2 | NP_003738.2 | 20 - 21 | 3152 | 67 |
XM_006716263.3 | XP_006716326.1 | 20 - 21 | 3858 | 67 | |
XM_011543845.2 | XP_011542147.1 | 20 - 21 | 3858 | 67 | |
XM_011543846.2 | XP_011542148.1 | 20 - 21 | 3858 | 67 | |
XM_011543847.2 | XP_011542149.1 | 18 - 19 | 3545 | 67 | |
GenBank mRNA | AB209339.1 | - | 19 - 20 | 2332 | 67 |
AF082556.1 | - | 20 - 21 | 3152 | 67 | |
AF082557.1 | - | 19 - 20 | 2448 | 67 | |
BC098394.1 | - | 20 - 21 | 3175 | 67 |