Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNTL5
Entrez Gene ID
168391
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
Gene Aliases
GALNACT19, GALNT15, GalNAc-T5L
Chromosome Location
Chr.7: 151956379 - 152019934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145292.3 NP_660335.2 2 - 3 514 159
NR_033169.1 3 - 4 618 159
XM_006715862.3 XP_006715925.1 3 - 4 589 159
XM_017011793.1 XP_016867282.1 2 - 3 1788 159
XM_017011795.1 XP_016867284.1 2 - 3 1889 159
XM_017011796.1 XP_016867285.1 2 - 3 1890 159
GenBank mRNA AF440400.1 - 3 - 4 553 159
AF440404.1 - 2 - 3 493 159
AK093091.1 - 3 - 4 571 159
AK315667.1 - 3 - 4 578 159
BC022021.2 - 2 - 3 467 159

View Full Product Details