The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003870.3 | NP_003861.1 | 13 - 14 | 1585 | 104 |
GenBank mRNA | AK307643.1 | - | 13 - 14 | 1502 | 104 |
BC020549.1 | - | 13 - 14 | 1529 | 104 | |
BC037425.1 | - | 13 - 14 | 1539 | 104 | |
BC064971.1 | - | 13 - 14 | 1535 | 104 | |
BC110811.1 | - | 13 - 14 | 1520 | 104 | |
BC139731.1 | - | 13 - 14 | 1530 | 104 | |
BC151834.1 | - | 13 - 14 | 1608 | 104 | |
D29640.1 | - | 13 - 14 | 1608 | 104 |