The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244710.1 | NP_001231639.1 | 15 - 16 | 1660 | 100 |
NM_002056.3 | NP_002047.2 | 14 - 15 | 1606 | 100 | |
XM_017003801.1 | XP_016859290.1 | 15 - 16 | 1624 | 100 | |
XM_017003802.1 | XP_016859291.1 | 14 - 15 | 1562 | 100 | |
GenBank mRNA | BC045641.1 | - | 14 - 15 | 1602 | 100 |
M90516.1 | - | 14 - 15 | 1545 | 100 |