The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152578.2 | NP_689791.1 | 3 - 4 | 614 | 73 |
GenBank mRNA | AK098602.1 | - | 3 - 4 | 599 | 73 |
AY211917.1 | - | 3 - 4 | 607 | 73 | |
BC034320.1 | - | 3 - 4 | 614 | 73 |