The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000461.4 | NP_000452.2 | 1 - 2 | 167 | 69 |
NM_001128177.1 | NP_001121649.1 | 1 - 2 | 167 | 69 | |
NM_001252634.1 | NP_001239563.1 | 1 - 2 | 167 | 69 | |
XM_006713317.3 | XP_006713380.1 | 1 - 2 | 372 | 69 | |
XM_006713318.3 | XP_006713381.1 | 1 - 2 | 372 | 69 | |
XM_011534047.2 | XP_011532349.1 | 1 - 2 | 372 | 69 | |
XM_011534050.2 | XP_011532352.1 | 1 - 2 | 372 | 69 | |
XM_017007109.1 | XP_016862598.1 | 1 - 2 | 372 | 69 | |
XM_017007113.1 | XP_016862602.1 | 1 - 2 | 372 | 69 | |
GenBank mRNA | AK096628.1 | - | 1 - 2 | 156 | 69 |
AY286465.1 | - | 1 - 2 | 93 | 69 | |
BP313943.1 | - | 1 - 2 | 156 | 69 | |
M26747.1 | - | 1 - 2 | 167 | 69 | |
X04707.1 | - | 1 - 2 | 167 | 69 |