Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHIT
Entrez Gene ID
2272
Gene Name
fragile histidine triad
Gene Aliases
AP3Aase, FRA3B
Chromosome Location
Chr.3: 59747278 - 61251474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166243.2 NP_001159715.1 3 - 4 263 87
NM_001320899.1 NP_001307828.1 3 - 4 263 87
NM_001320900.1 NP_001307829.1 4 - 5 357 87
NM_002012.3 NP_002003.1 3 - 4 263 87
XM_017005880.1 XP_016861369.1 3 - 4 261 87
XM_017005882.1 XP_016861371.1 3 - 4 261 87
XM_017005883.1 XP_016861372.1 3 - 4 261 87
XM_017005884.1 XP_016861373.1 3 - 4 261 87
GenBank mRNA AK289824.1 - 3 - 4 261 87
BC032336.1 - 3 - 4 215 87
DA239143.1 - 3 - 4 261 87
DB477833.1 - 3 - 4 263 87
U46922.1 - 3 - 4 252 87

View Full Product Details