The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177599.1 | NP_001171070.1 | 8 - 9 | 950 | 133 |
NM_003848.3 | NP_003839.2 | 8 - 9 | 950 | 133 | |
XM_017007415.1 | XP_016862904.1 | 8 - 9 | 946 | 133 | |
XM_017007416.1 | XP_016862905.1 | 8 - 9 | 946 | 133 | |
XM_017007417.1 | XP_016862906.1 | 8 - 9 | 946 | 133 | |
XM_017007418.1 | XP_016862907.1 | 8 - 9 | 946 | 133 | |
XM_017007419.1 | XP_016862908.1 | 8 - 9 | 946 | 133 | |
XM_017007420.1 | XP_016862909.1 | 8 - 9 | 946 | 133 | |
GenBank mRNA | AF058954.1 | - | 7 - 8 | 838 | 133 |
AF131748.1 | - | 2 - 3 | 223 | 133 | |
AK294598.1 | - | 8 - 9 | 943 | 133 | |
AK310527.1 | - | 8 - 9 | 946 | 133 | |
BC007716.2 | - | 8 - 9 | 913 | 133 | |
BC019868.1 | - | 5 - 6 | 589 | 133 | |
BC035149.1 | - | 7 - 8 | 712 | 133 | |
BC047024.1 | - | 9 - 10 | 920 | 133 | |
BC068602.1 | - | 8 - 9 | 951 | 133 |