The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002800.4 | NP_002791.1 | 4 - 5 | 456 | 78 |
GenBank mRNA | AK303118.1 | - | 3 - 4 | 327 | 78 |
BC065513.1 | - | 4 - 5 | 411 | 78 | |
BI832878.1 | - | 4 - 5 | 438 | 78 | |
BM998687.1 | - | 2 - 3 | 78 | ||
CR541656.1 | - | 4 - 5 | 387 | 78 | |
S75169.1 | - | 3 - 4 | 366 | 78 | |
U01025.1 | - | 3 - 4 | 366 | 78 | |
X62741.1 | - | 4 - 5 | 402 | 78 |