Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCRA
Entrez Gene ID
171558
Gene Name
pre T-cell antigen receptor alpha
Gene Aliases
PT-ALPHA, PTA
Chromosome Location
Chr.6: 42915989 - 42925838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243170.1 NP_001230099.1 2 - 3 184 117
GenBank mRNA AF101436.1 - 2 - 3 103 117

View Full Product Details