Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
Chromosome Location
Chr.15: 74630558 - 74696045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142443.1 NP_001135915.1 6 - 7 971 73
NM_001142444.1 NP_001135916.1 4 - 5 756 73
NM_025083.3 NP_079359.2 3 - 4 667 73
GenBank mRNA AF193058.1 - 4 - 5 704 73
AK024781.1 - 4 - 5 696 73
AK056339.1 - 3 - 4 667 73
AK225099.1 - 4 - 5 696 73
BC011534.1 - 3 - 4 641 73
BC021271.2 - 6 - 7 945 73

View Full Product Details