Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
Chromosome Location
Chr.15: 74630558 - 74696045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142443.1 NP_001135915.1 8 - 9 1468 84
NM_001142444.1 NP_001135916.1 6 - 7 1253 84
NM_025083.3 NP_079359.2 5 - 6 1164 84
XM_011522092.2 XP_011520394.1 3 - 4 574 84
GenBank mRNA AF193058.1 - 6 - 7 1201 84
AK024781.1 - 6 - 7 1193 84
AK056339.1 - 5 - 6 1164 84
AK225099.1 - 6 - 7 1193 84
BC011534.1 - 5 - 6 1138 84
BC021271.2 - 8 - 9 1442 84

View Full Product Details