Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
Chromosome Location
Chr.15: 74337762 - 74367740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000781.2 NP_000772.2 3 - 4 777 94
NM_001099773.1 NP_001093243.1 3 - 4 851 94
GenBank mRNA AK056794.1 - 3 - 4 846 94
AK292300.1 - 3 - 4 683 94
AK292301.1 - 2 - 3 641 94
AK293211.1 - 4 - 5 642 94
AK300178.1 - 3 - 4 593 94
BC032329.1 - 3 - 4 666 94
KU178011.1 - 3 - 4 622 94
M14565.1 - 3 - 4 666 94

View Full Product Details