Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SCN1A
- Entrez Gene ID
-
6323
- Gene Name
- sodium voltage-gated channel alpha subunit 1
- Gene Aliases
-
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
- Chromosome Location
-
Chr.2: 165989160 - 166149216 on Build GRCh38
View Full Product Details