Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN1A
Entrez Gene ID
6323
Gene Name
sodium voltage-gated channel alpha subunit 1
Gene Aliases
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
Chromosome Location
Chr.2: 165989160 - 166149216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165963.1 NP_001159435.1 6 - 7 981 108
NM_001165964.1 NP_001159436.1 6 - 7 981 108
NM_001202435.1 NP_001189364.1 8 - 9 1190 108
NM_006920.4 NP_008851.3 6 - 7 981 108
XM_011511602.2 XP_011509904.1 7 - 8 1257 108
XM_011511604.2 XP_011509906.1 7 - 8 1150 108
XM_011511605.2 XP_011509907.1 8 - 9 1357 108
XM_011511606.2 XP_011509908.1 8 - 9 1349 108
XM_017004644.1 XP_016860133.1 9 - 10 1436 108
XM_017004645.1 XP_016860134.1 8 - 9 1357 108
XM_017004646.1 XP_016860135.1 7 - 8 1271 108
XM_017004647.1 XP_016860136.1 9 - 10 1443 108
XM_017004648.1 XP_016860137.1 8 - 9 1220 108
XM_017004649.1 XP_016860138.1 7 - 8 1182 108
XM_017004650.1 XP_016860139.1 7 - 8 1259 108
XM_017004651.1 XP_016860140.1 8 - 9 1212 108
XM_017004652.1 XP_016860141.1 7 - 8 1252 108
XM_017004653.1 XP_016860142.1 8 - 9 1362 108
XM_017004654.1 XP_016860143.1 8 - 9 1373 108
GenBank mRNA AB093548.1 - 6 - 7 963 108
AB093549.1 - 6 - 7 963 108
AB098335.1 - 6 - 7 963 108
AF225985.1 - 6 - 7 981 108
AY043484.1 - 6 - 7 963 108

View Full Product Details