The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001998.2 | NP_001001998.1 | 22 - 23 | 2569 | 143 |
NM_002685.3 | NP_002676.1 | 21 - 22 | 2494 | 143 | |
XM_005263475.2 | XP_005263532.2 | 22 - 23 | 2543 | 143 | |
XM_017001494.1 | XP_016856983.1 | 21 - 22 | 2269 | 143 | |
XM_017001495.1 | XP_016856984.1 | 21 - 22 | 2254 | 143 | |
XM_017001496.1 | XP_016856985.1 | 22 - 23 | 2614 | 143 | |
XM_017001497.1 | XP_016856986.1 | 22 - 23 | 2599 | 143 | |
GenBank mRNA | AB209236.1 | - | 18 - 19 | 4101 | 143 |
AK123365.1 | - | 10 - 11 | 1742 | 143 | |
AK296559.1 | - | 20 - 21 | 3069 | 143 | |
BC009908.2 | - | 18 - 19 | 1891 | 143 | |
BC039901.1 | - | 22 - 23 | 2510 | 143 | |
BC073788.1 | - | 22 - 23 | 2545 | 143 | |
CA307576.1 | - | 3 - 4 | 143 | ||
L01457.1 | - | 21 - 22 | 2453 | 143 | |
X66113.1 | - | 22 - 23 | 2527 | 143 |