Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNE1
Entrez Gene ID
3753
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1
Gene Aliases
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
Chromosome Location
Chr.21: 34446688 - 34512275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000219.5 NP_000210.2 4 - 4 1055 89
NM_001127668.3 NP_001121140.1 3 - 3 667 89
NM_001127669.3 NP_001121141.1 3 - 3 639 89
NM_001127670.3 NP_001121142.1 2 - 2 904 89
NM_001270402.2 NP_001257331.1 3 - 3 830 89
NM_001270403.2 NP_001257332.1 3 - 3 778 89
NM_001270404.2 NP_001257333.1 2 - 2 719 89
NM_001270405.2 NP_001257334.1 3 - 3 631 89
XM_017028342.1 XP_016883831.1 2 - 2 2407 89
GenBank mRNA AK304336.1 - 3 - 3 723 89
AK311384.1 - 4 - 4 242 89
BC036452.1 - 3 - 3 641 89
BC046224.1 - 4 - 4 1055 89

Gene Symbol
KCNE1B
Entrez Gene ID
102723475
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1B
Gene Aliases
-
Chromosome Location
Chr.21: 7816677 - 7829632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723946.2 XP_006724009.1 2 - 2 860 89
XM_006723949.2 XP_006724012.1 3 - 3 707 89

View Full Product Details