The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145860.1 | NP_001139332.1 | 7 - 8 | 1108 | 65 |
NM_001145861.1 | NP_001139333.1 | 7 - 8 | 1048 | 65 | |
NM_015029.2 | NP_055844.2 | 7 - 8 | 1036 | 65 | |
XM_011516801.2 | XP_011515103.1 | 7 - 8 | 1108 | 65 | |
GenBank mRNA | AK000837.1 | - | 1 - 2 | 102 | 65 |
AK291434.1 | - | 7 - 8 | 1105 | 65 | |
BC011529.1 | - | 7 - 8 | 1036 | 65 | |
D31765.1 | - | 4 - 5 | 647 | 65 |