The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002837.2 | NP_001002837.1 | 2 - 3 | 260 | 62 |
GenBank mRNA | AK126610.1 | - | 2 - 3 | 189 | 62 |