Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D10A
Entrez Gene ID
83874
Gene Name
TBC1 domain family member 10A
Gene Aliases
EPI64, TBC1D10, dJ130H16.1, dJ130H16.2
Chromosome Location
Chr.22: 30291990 - 30326966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204240.1 NP_001191169.1 8 - 9 1158 65
NM_031937.2 NP_114143.1 8 - 9 1137 65
GenBank mRNA AB449894.1 - 8 - 9 1052 65
AK074656.1 - 8 - 9 1103 65
AK093231.1 - 5 - 6 2272 65
AK127939.1 - 8 - 9 1093 65
AK131086.1 - 4 - 5 4702 65
AK309455.1 - 8 - 9 903 65
BC022201.1 - 8 - 9 1086 65
BC136815.1 - 8 - 9 1093 65
BC142940.1 - 8 - 9 1109 65
BC150187.1 - 8 - 9 1057 65
BC150214.1 - 8 - 9 1062 65
BC171775.1 - 8 - 9 1093 65
CT841514.1 - 8 - 9 1058 65

View Full Product Details