The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002877.1 | NP_001002877.1 | 7 - 8 | 805 | 91 |
NM_001002878.1 | NP_001002878.1 | 7 - 8 | 936 | 91 | |
NM_001002879.1 | NP_001002879.1 | 6 - 7 | 820 | 91 | |
NM_003678.4 | NP_003669.4 | 6 - 7 | 689 | 91 | |
XM_005261797.1 | XP_005261854.1 | 6 - 7 | 702 | 91 | |
XM_005261798.1 | XP_005261855.1 | 7 - 8 | 818 | 91 | |
XM_005261799.1 | XP_005261856.1 | 6 - 7 | 845 | 91 | |
XM_005261801.3 | XP_005261858.1 | 6 - 7 | 643 | 91 | |
XM_006724347.1 | XP_006724410.1 | 7 - 8 | 932 | 91 | |
XM_017029027.1 | XP_016884516.1 | 7 - 8 | 1304 | 91 | |
GenBank mRNA | AB023200.1 | - | 6 - 7 | 656 | 91 |
AK025385.1 | - | 6 - 7 | 639 | 91 | |
AK098709.1 | - | 3 - 4 | 306 | 91 | |
AK122673.1 | - | 7 - 8 | 1312 | 91 | |
AK225169.1 | - | 5 - 6 | 598 | 91 | |
BC003615.2 | - | 6 - 7 | 629 | 91 | |
BX381026.2 | - | 7 - 8 | 771 | 91 | |
BX404329.2 | - | 6 - 7 | 693 | 91 | |
CR456542.1 | - | 5 - 6 | 623 | 91 |