The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006190.4 | NP_006181.1 | 6 - 7 | 693 | 74 |
NR_033915.1 | 6 - 7 | 693 | 74 | ||
XM_006712555.3 | XP_006712618.1 | 6 - 7 | 640 | 74 | |
XM_011511252.2 | XP_011509554.1 | 7 - 8 | 755 | 74 | |
XM_011511253.2 | XP_011509555.1 | 7 - 8 | 627 | 74 | |
GenBank mRNA | AK302614.1 | - | 6 - 7 | 665 | 74 |
AK316433.1 | - | 6 - 7 | 594 | 74 | |
BC014834.1 | - | 6 - 7 | 636 | 74 | |
BM470184.1 | - | - | 680 | 74 | |
U40268.2 | - | 6 - 7 | 608 | 74 |