The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114090.1 | NP_001107562.1 | 7 - 8 | 1220 | 62 |
NM_019850.2 | NP_062824.2 | 9 - 10 | 1592 | 62 | |
XM_011510923.2 | XP_011509225.1 | 9 - 10 | 1558 | 62 | |
XM_011510924.1 | XP_011509226.1 | 5 - 6 | 591 | 62 | |
XM_011510925.1 | XP_011509227.1 | 5 - 6 | 595 | 62 | |
GenBank mRNA | AJ238899.1 | - | 6 - 7 | 62 | |
AK293203.1 | - | 5 - 6 | 62 | ||
AK297390.1 | - | 7 - 8 | 62 | ||
AK298315.1 | - | 9 - 10 | 1544 | 62 | |
AK300953.1 | - | 10 - 11 | 62 | ||
BC031573.1 | - | 9 - 10 | 1579 | 62 | |
BX431262.1 | - | 4 - 5 | 465 | 62 |