Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MATN4
Entrez Gene ID
8785
Gene Name
matrilin 4
Gene Aliases
-
Chromosome Location
Chr.20: 45293446 - 45308566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003833.4 NP_003824.2 6 - 7 1130 59
NM_030590.3 NP_085080.1 4 - 5 949 59
XM_005260597.1 XP_005260654.1 5 - 6 1311 59
XM_017028113.1 XP_016883602.1 6 - 7 1130 59
XM_017028114.1 XP_016883603.1 4 - 5 1188 59
GenBank mRNA AJ007581.1 - 4 - 5 894 59
AK074495.1 - 2 - 3 328 59
AK074593.1 - 7 - 8 1130 59
AK074595.1 - 4 - 5 866 59
AK074597.1 - 4 - 5 951 59
BC131763.1 - 4 - 5 898 59
BC141811.1 - 4 - 5 937 59
BC143625.1 - 4 - 5 857 59
BC151219.1 - 5 - 6 1025 59

View Full Product Details